6-Chloro-8-fluoroimidazo[1,2-a]pyridine 98%
Dodavatel: Apollo Scientific
PC902162-1G
PC902162-5G
APOSPC902162-1GEA
1210
CZK
APOSPC902162-1G
APOSPC902162-5G
6-Chloro-8-fluoroimidazo[1,2-a]pyridine 98%
6-Chloro-8-fluoroimidazo[1,2-a]pyridine
Vzorec:
C₇H₄ClFN₂ MW: 170,5700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10699670 CAS číslo: 1033202-10-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...