4,6-Difluoro-2H-benzo[d][1,2,3]triazole 98%
Dodavatel: Apollo Scientific
PC901968-1G
APOSPC901968-1GEA
5230
CZK
APOSPC901968-1G
4,6-Difluoro-2H-benzo[d][1,2,3]triazole 98%
4,6-Difluoro-2H-benzo[d][1,2,3]triazole
Vzorec:
C₆H₃F₂N₃ MW: 155,10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16295089 CAS číslo: 2208-25-5 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...