Tisk…

5,7-Difluoroquinoxaline 97%

Dodavatel: Apollo Scientific

PC901966-1G PC901966-5G PC901966-25G
APOSPC901966-1GEA 3540 CZK
APOSPC901966-1G APOSPC901966-5G APOSPC901966-25G
5,7-Difluoroquinoxaline 97%
5,7-Difluoroquinoxaline
Vzorec: C₈H₄F₂N₂
Storage Temperature: Ambient
MDL Number: MFCD15143566
CAS číslo: 1215205-81-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR