Tisk…
3-Chloro-2-fluoro-N-methylaniline hydrochloride 98%
Dodavatel: Apollo Scientific
PC901919-1G PC901919-5G
APOSPC901919-1GEA 7250 CZK
APOSPC901919-1G APOSPC901919-5G
3-Chloro-2-fluoro-N-methylaniline hydrochloride 98%
3-Chloro-2-fluoro-N-methylaniline hydrochloride
Vzorec: C₇H₈Cl₂FN
Storage Temperature: Ambient
MDL Number: MFCD12913956
CAS číslo: 1187386-17-9