8-Fluoro-5-nitroquinoline 95%
Dodavatel: Apollo Scientific
PC901843-1G
PC901843-5G
APOSPC901843-1GEA
6550
CZK
APOSPC901843-1G
APOSPC901843-5G
8-Fluoro-5-nitroquinoline 95%
8-Fluoro-5-nitroquinoline
Vzorec:
C₉H₅FN₂O₂ MW: 192,1466 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09048716 CAS číslo: 94832-39-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...