Tisk…
2-(2-Fluorophenyl)-2-methylpropionic acid 98%
Dodavatel: Apollo Scientific
Synonyms: 2-(2-Fluorophenyl)-2-methylpropanoic acid
PC901552-1G PC901552-5G
APOSPC901552-1GEA 6090 CZK
APOSPC901552-1G APOSPC901552-5G
2-(2-Fluorophenyl)-2-methylpropionic acid 98%
2-(2-Fluorophenyl)-2-methylpropionic acid
Vzorec: C₁₀H₁₁FO₂
MW: 182,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11036900
CAS číslo: 870849-49-3