Tisk…

N,N-Diethyl 2-fluorobenzamide 98%

Dodavatel: Apollo Scientific

PC901459-1G PC901459-25G PC901459-5G
APOSPC901459-1GEA 3540 CZK
APOSPC901459-1G APOSPC901459-25G APOSPC901459-5G
N,N-Diethyl 2-fluorobenzamide 98%
N,N-Diethyl 2-fluorobenzamide
Vzorec: C₁₁H₁₄FNO
Storage Temperature: Ambient
MDL Number: MFCD00460287
CAS číslo: 10345-76-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR