Tisk…

5-Fluoro-1,3-dihydrobenzoimidazol-2-one 96%

Dodavatel: Apollo Scientific

Synonyms: 5-Fluoro-1H-benzo[d]imidazol-2(3H)-one

PC901184-5G
APOSPC901184-5GEA 6680 CZK
APOSPC901184-5G
5-Fluoro-1,3-dihydrobenzoimidazol-2-one 96%
5-Fluoro-1,3-dihydrobenzoimidazol-2-one
Vzorec: C₇H₅FN₂O
MW: 152,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663528
CAS číslo: 1544-75-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR