Tisk…

5-Fluoro-2-methylpyrimidin-4(3H)-one 95%

Dodavatel: Apollo Scientific

Synonyms: 5-Fluoro-2-methylpyrimidin-4-ol

PC901130-1G PC901130-250MG
APOSPC901130-1GEA 4860 CZK
APOSPC901130-1G APOSPC901130-250MG
5-Fluoro-2-methylpyrimidin-4(3H)-one 95%
5-Fluoro-2-methylpyrimidin-4(3H)-one
Vzorec: C₅H₅FN₂O
MW: 128,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16988103
CAS číslo: 1480-91-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR