Tisk…

7-(Trifluoromethyl)quinoline-3-carboxylic acid 97%

Dodavatel: Apollo Scientific

PC901003-1G PC901003-250MG
APOSPC901003-1GEA 5790 CZK
APOSPC901003-1G APOSPC901003-250MG
7-(Trifluoromethyl)quinoline-3-carboxylic acid 97%
7-(Trifluoromethyl)quinoline-3-carboxylic acid
Vzorec: C₁₁H₆F₃NO₂
MW: 241,1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042923
CAS číslo: 71082-51-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR