2-Difluoromethyl-2H-pyrazole-3-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC900917-1G
PC900917-25G
PC900917-5G
APOSPC900917-1GEA
10850
CZK
APOSPC900917-1G
APOSPC900917-25G
APOSPC900917-5G
2-Difluoromethyl-2H-pyrazole-3-carboxylic acid
2-Difluoromethyl-2H-pyrazole-3-carboxylic acid
Vzorec:
C₅H₄F₂N₂O₂ MW: 162,1 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03420213 CAS číslo: 925199-97-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...