Tisk…

2-(4-Fluorophenyl)piperidine 95%

Dodavatel: Apollo Scientific

PC900699-1G PC900699-5G
APOSPC900699-1GEA 6420 CZK
APOSPC900699-1G APOSPC900699-5G
2-(4-Fluorophenyl)piperidine 95%
2-(4-Fluorophenyl)piperidine
Vzorec: C₁₁H₁₄FN
Storage Temperature: Ambient
MDL Number: MFCD02663556
CAS číslo: 383128-03-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR