5-Fluoro-2-pyrrolidin-1-ylpyridine 96%
Dodavatel: Apollo Scientific
PC900592-1G
PC900592-5G
APOSPC900592-1GEA
4770
CZK
APOSPC900592-1G
APOSPC900592-5G
5-Fluoro-2-pyrrolidin-1-ylpyridine 96%
5-Fluoro-2-pyrrolidin-1-ylpyridine
Vzorec:
C₉H₁₁FN₂ MW: 166,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18089240 CAS číslo: 1287217-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...