Tisk…

3-[3-(Trifluoromethyl)phenyl]propanenitrile 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-(3-(Trifluoromethyl)phenyl)propanenitrile

PC8987-1G PC8987-250MG
APOSPC8987-1GEA 6720 CZK
APOSPC8987-1G APOSPC8987-250MG
3-[3-(Trifluoromethyl)phenyl]propanenitrile 95%
3-[3-(Trifluoromethyl)phenyl]propanenitrile
Vzorec: C₁₀H₈F₃N
MW: 199,18 g/mol
Bod varu: 86 °C/0,3mmHg
Storage Temperature: Ambient
MDL Number: MFCD04973009
CAS číslo: 95096-06-3
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR