Trimethylsilyl trifluoroacetate 96%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Trifluoroacetic Acid Trimethylsilyl Ester
PC8977-25G
APOSPC8977-25GEA
2780
CZK
APOSPC8977-25G
Trimethylsilyl trifluoroacetate 96%
Trimethylsilyl trifluoroacetate
Reagent for ring opening of cyclopropyl ketones.
Vzorec:
C₅H₉F₃O₂Si MW: 186,21 g/mol Bod varu: 88…90 °C Hustota: 1,078 g/cm³ (25 °C) Bod vzplanutí: 13 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000413 CAS číslo: 400-53-3 UN: 2924 ADR: 3+8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...