6-(Trifluoromethyl)picolinic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-(Trifluoromethyl)-2-pyridinecarboxylic acid, 6-(Trifluoromethyl)picolinic acid, 2-(Trifluoromethyl)-6-pyridinecarboxylic acid, 6-(Trifluoromethyl)pyridine-2-carboxylic acid
PC8969-1G
PC8969-250MG
PC8969-5G
PC8969-25G
APOSPC8969-1GEA
622
CZK
APOSPC8969-1G
APOSPC8969-250MG
APOSPC8969-5G
APOSPC8969-25G
6-(Trifluoromethyl)picolinic acid 98%
6-(Trifluoromethyl)picolinic acid
Vzorec:
C₇H₄F₃NO₂ MW: 191,11 g/mol Bod tání: 154…158 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02180819 CAS číslo: 131747-42-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...