Tisk…

2-(4-Fluorophenyl)propan-2-amine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(4-Fluorophenyl)-1-methylethylamine, alpha, alpha-Dimethyl-4-fluorobenzylamine

PC8957-1G PC8957-10G PC8957-5G
APOSPC8957-1GEA 2980 CZK
APOSPC8957-1G APOSPC8957-10G APOSPC8957-5G
2-(4-Fluorophenyl)propan-2-amine
2-(4-Fluorophenyl)propan-2-amine
Vzorec: C₉H₁₂FN
Bod varu: 40 °C/5,25mmHg
Bod vzplanutí: 92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07777176
CAS číslo: 17797-10-3
UN: 3259
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR