Tisk…

Ethyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylate

Dodavatel: Apollo Scientific

PC8922-1G PC8922-250MG
APOSPC8922-1GEA 2480 CZK
APOSPC8922-1G APOSPC8922-250MG
Ethyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylate
Ethyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylate
Vzorec: C₁₃H₁₀F₃NO₂S
MW: 301,29 g/mol
Bod tání: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00142038
CAS číslo: 132089-39-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR