Perfluoro-3,6-dioxaoctane-1,8-dioic acid 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC8914-5G
PC8914-1G
APOSPC8914-5GEA
1870
CZK
APOSPC8914-5G
APOSPC8914-1G
Perfluoro-3,6-dioxaoctane-1,8-dioic acid 98%
Perfluoro-3,6-dioxaoctane-1,8-dioic acid
Vzorec:
C₆H₂F₈O₆ Bod varu: 115 °C/0,8mmHg Storage Temperature: Ambient |
MDL Number:
MFCD03094537 CAS číslo: 55621-21-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...