Tisk…

3,5-Difluorophenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (3,5-Difluorophenyl)metaboric acid, Dihydroxy(3,5-difluorophenyl)borane, 3,5-Difluorobenzeneboronic acid

PC8911-100G PC8911-25G PC8911-5G
APOSPC8911-100GEA 2820 CZK
APOSPC8911-100G APOSPC8911-25G APOSPC8911-5G
3,5-Difluorophenylboronic acid 98%
3,5-Difluorophenylboronic acid
Vzorec: C₆H₅BF₂O₂
MW: 157.91
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD01318138
CAS číslo: 156545-07-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR