a-Tosyl(4-fluorobenzyl)isocyanide 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(4-Fluorophenyl)(isocyano)methyl 4-methylphenyl sulphone, a-Tosyl-(4-fluorobenzyl) isocyanide
PC8908-1G
PC8908-10G
PC8908-5G
APOSPC8908-1GEA
4070
CZK
APOSPC8908-1G
APOSPC8908-10G
APOSPC8908-5G
a-Tosyl(4-fluorobenzyl)isocyanide 98%
a-Tosyl(4-fluorobenzyl)isocyanide
Vzorec:
C₁₅H₁₂FNO₂S MW: 289,32 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03787932 CAS číslo: 165806-95-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...