Tisk…

3-(Difluoromethoxy)benzylamine 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC8902-1G PC8902-5G PC8902-10G
APOSPC8902-1GEA 1080 CZK
APOSPC8902-1G APOSPC8902-5G APOSPC8902-10G
3-(Difluoromethoxy)benzylamine 98%
3-(Difluoromethoxy)benzylamine
Vzorec: C₈H₉F₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD00236230
CAS číslo: 244022-71-7
UN: 2735
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR