Tisk…

2,5-Dibromo-3,4,6-trifluoroaniline

Dodavatel: Apollo Scientific
Varování

PC8860-1G PC8860-5G
APOSPC8860-1GEA 2730 CZK
APOSPC8860-1G APOSPC8860-5G
2,5-Dibromo-3,4,6-trifluoroaniline
2,5-Dibromo-3,4,6-trifluoroaniline
Vzorec: C₆H₂Br₂F₃N
Storage Temperature: Ambient
MDL Number: MFCD01862003
CAS číslo: 232267-32-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR