Tisk…

2-Fluoro-4-(phenylsulphonyl)phenylhydrazine

Dodavatel: Apollo Scientific

PC8858-1G PC8858-5G
APOSPC8858-1GEA 4730 CZK
APOSPC8858-1G APOSPC8858-5G
2-Fluoro-4-(phenylsulphonyl)phenylhydrazine
2-Fluoro-4-(phenylsulphonyl)phenylhydrazine
Vzorec: C₁₂H₁₁FN₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD09800751
CAS číslo: 1000339-96-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR