Tisk…

4'-Bromo-3',5'-difluoroacetanilide

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: N-(4-Bromo-3,5-difluorophenyl)acetamide

PC8844-100G PC8844-25G PC8844-5G
APOSPC8844-100GEA 19480 CZK
APOSPC8844-100G APOSPC8844-25G APOSPC8844-5G
4'-Bromo-3',5'-difluoroacetanilide
4'-Bromo-3',5'-difluoroacetanilide
Vzorec: C₈H₆BrF₂NO
Storage Temperature: Ambient
MDL Number: MFCD04038226
CAS číslo: 541539-64-4
UN: 2811
ADR: 6.1 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR