4-(Chlorodifluoromethyl)benzotrifluoride
Dodavatel: Apollo Scientific
|
Varování
|
PC8829-1G
PC8829-5G
APOSPC8829-1GEA
11540
CZK
APOSPC8829-1G
APOSPC8829-5G
4-(Chlorodifluoromethyl)benzotrifluoride
4-(Chlorodifluoromethyl)benzotrifluoride
Vzorec:
C₈H₄ClF₅ Storage Temperature: Ambient |
CAS číslo:
13947-94-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...