Tisk…

4-(4-Chloro-3-fluorophenyl)benzaldehyde

Dodavatel: Apollo Scientific

Synonyms: 4'-Chloro-3'-fluoro-[1,1'-biphenyl]-4-carboxaldehyde, 4'-Chloro-3'-fluoro[1,1'-biphenyl]-4-carbaldehyde

PC8777-10G PC8777-1G PC8777-5G
APOSPC8777-10GEA 47940 CZK
APOSPC8777-10G APOSPC8777-1G APOSPC8777-5G
4-(4-Chloro-3-fluorophenyl)benzaldehyde
4-(4-Chloro-3-fluorophenyl)benzaldehyde
Vzorec: C₁₃H₈ClFO
Bod tání: 112…115 °C
Storage Temperature: Ambient
MDL Number: MFCD06201223
CAS číslo: 842140-45-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR