Tisk…

2-methyl-4-(trifluoromethoxy)phenol 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Methyl-4-(trifluoromethoxy)phenol

PC8770-1G PC8770-250MG PC8770-5G
APOSPC8770-1GEA 705 CZK
APOSPC8770-1G APOSPC8770-250MG APOSPC8770-5G
2-methyl-4-(trifluoromethoxy)phenol 97%
2-methyl-4-(trifluoromethoxy)phenol
Vzorec: C₈H₇F₃O₂
MW: 192.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972927
CAS číslo: 129676-67-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR