3-(Trifluoromethoxy)cinnamic acid 96%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3-(3-(Trifluoromethoxy)phenyl)acrylic acid
PC8754-25G
PC8754-5G
PC8754-100G
APOSPC8754-25GEA
1740
CZK
APOSPC8754-25G
APOSPC8754-5G
APOSPC8754-100G
3-(Trifluoromethoxy)cinnamic acid 96%
3-(Trifluoromethoxy)cinnamic acid
Vzorec:
C₁₀H₇F₃O₃ MW: 232,16 g/mol Bod tání: 92…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066337 CAS číslo: 168833-80-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...