5-Bromo-2-chloro-1,3-difluorobenzene 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Chloro-3,5-difluorobromobenzene
PC8717-25G
PC8717-5G
PC8717-1G
PC8717-100G
APOSPC8717-25GEA
1330
CZK
APOSPC8717-25G
APOSPC8717-5G
APOSPC8717-1G
APOSPC8717-100G
5-Bromo-2-chloro-1,3-difluorobenzene 98%
5-Bromo-2-chloro-1,3-difluorobenzene
Vzorec:
C₆H₂BrClF₂ MW: 227,4300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00143420 CAS číslo: 176673-72-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...