3-Fluoroisatoic anhydride
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
8-Fluoro-1H-benzo[d][1,3]oxazine-2,4-dione
PC8703-1G
PC8703-250MG
PC8703-5G
APOSPC8703-1GEA
1780
CZK
APOSPC8703-1G
APOSPC8703-250MG
APOSPC8703-5G
3-Fluoroisatoic anhydride
3-Fluoroisatoic anhydride
Vzorec:
C₈H₄FNO₃ MW: 181,1207 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06796569 CAS číslo: 174463-53-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...