4-Fluoroisatoic anhydride
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
7-Fluoro-1H-benzo[d][1,3]oxazine-2,4-dione
PC8702-1G
PC8702-25G
PC8702-5G
APOSPC8702-1GEA
622
CZK
APOSPC8702-1G
APOSPC8702-25G
APOSPC8702-5G
4-Fluoroisatoic anhydride
4-Fluoroisatoic anhydride
Vzorec:
C₈H₄FNO₃ Bod tání: 231…232 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03839826 CAS číslo: 321-50-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...