Tisk…

5-Fluoroisatoic anhydride

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Fluoro-2H-benzo[d][1,3]oxazine-2,4(1H)-dione

PC8701-1G PC8701-25G PC8701-5G
APOSPC8701-1GEA 622 CZK
APOSPC8701-1G APOSPC8701-25G APOSPC8701-5G
5-Fluoroisatoic anhydride
5-Fluoroisatoic anhydride
Vzorec: C₈H₄FNO₃
MW: 181,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03300719
CAS číslo: 321-69-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR