3-[(4-Fluorobenzyl)oxy]-2-thiophenecarboxylic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC8641-10G
PC8641-1G
PC8641-5G
APOSPC8641-10GEA
32040
CZK
APOSPC8641-10G
APOSPC8641-1G
APOSPC8641-5G
3-[(4-Fluorobenzyl)oxy]-2-thiophenecarboxylic acid 97%
3-[(4-Fluorobenzyl)oxy]-2-thiophenecarboxylic acid
Vzorec:
C₁₂H₉FO₃S Bod tání: 118…120 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00173430 CAS číslo: 303147-78-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...