4'-Chloro-3'-fluoro[1,1'-biphenyl]-4-carboxylic acid
Dodavatel: Apollo Scientific
PC8639-500MG
PC8639-1G
APOSPC8639-500MGEA
0
CZK
APOSPC8639-500MG
APOSPC8639-1G
4'-Chloro-3'-fluoro[1,1'-biphenyl]-4-carboxylic acid
4'-Chloro-3'-fluoro[1,1'-biphenyl]-4-carboxylic acid
Vzorec:
C₁₃H₈ClFO₂ MW: 250,66 g/mol Bod tání: 270…272 °C Storage Temperature: Ambient |
MDL Number:
MFCD06201432 CAS číslo: 844878-86-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...