Tisk…

1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodononane 97%

Dodavatel: Apollo Scientific

Synonyms: 2-Iodo-1H,1H,1H,2H,3H,3H-perfluorononane

PC8627-1G PC8627-5G
APOSPC8627-1GEA 2420 CZK
APOSPC8627-1G APOSPC8627-5G
1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodononane 97%
1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodononane
Vzorec: C₉H₆F₁₃I
Storage Temperature: Ambient
MDL Number: MFCD00156018
CAS číslo: 38550-34-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR