2-Cyano-N-[4-(trifluoromethyl)benzyl]acetamide
Dodavatel: Apollo Scientific
PC8620-10G
PC8620-1G
PC8620-5G
APOSPC8620-10GEA
7210
CZK
APOSPC8620-10G
APOSPC8620-1G
APOSPC8620-5G
2-Cyano-N-[4-(trifluoromethyl)benzyl]acetamide
2-Cyano-N-[4-(trifluoromethyl)benzyl]acetamide
Vzorec:
C₁₁H₉F₃N₂O Bod tání: 125…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD08689696 CAS číslo: 566926-08-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...