6-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC8616-1G
PC8616-250MG
PC8616-5G
APOSPC8616-1GEA
4900
CZK
APOSPC8616-1G
APOSPC8616-250MG
APOSPC8616-5G
6-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid
6-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid
Vzorec:
C₁₀H₅F₃O₂S Bod tání: 198…201 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07371544 CAS číslo: 142329-22-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...