Methyl-5-(trifluoromethyl)benzo[b]thiophene-2-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 5-(trifluoromethyl)benzo[b]thiophene-2-carboxylate
PC8598-100MG
PC8598-1G
APOSPC8598-100MGEA
622
CZK
APOSPC8598-100MG
APOSPC8598-1G
Methyl-5-(trifluoromethyl)benzo[b]thiophene-2-carboxylate
Methyl-5-(trifluoromethyl)benzo[b]thiophene-2-carboxylate
Vzorec:
C₁₁H₇F₃O₂S MW: 260,2323 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03012932 CAS číslo: 146137-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...