Tisk…

Trifluoro(pentafluoroethoxy)ethylene

Dodavatel: Apollo Scientific
Varování

Synonyms: Pentafluoroethyl trifluorovinyl ether

PC8568-25G PC8568-5G
APOSPC8568-25GEA 12240 CZK
APOSPC8568-25G APOSPC8568-5G
Trifluoro(pentafluoroethoxy)ethylene
Trifluoro(pentafluoroethoxy)ethylene
Vzorec: C₄F₈O
MW: 216,03 g/mol
Bod varu: 7,4 °C
Hustota: 1,445 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02101232
CAS číslo: 10493-43-3
UN: 3154
ADR: 2.1,

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR