4-Bromo-2-fluorobenzoyl chloride 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-Bromo-2-fluorobenzene-1-carbonyl chloride
PC8531-25G
PC8531-5G
PC8531-100G
APOSPC8531-25GEA
2400
CZK
APOSPC8531-25G
APOSPC8531-5G
APOSPC8531-100G
4-Bromo-2-fluorobenzoyl chloride 98%
4-Bromo-2-fluorobenzoyl chloride
Vzorec:
C₇H₃BrClFO Bod varu: 86…88 °C/15mmHg Bod tání: 24…25 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411584 CAS číslo: 151982-51-3 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...