Tisk…

3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}-5-methyl-4-isothiazolecarbonitrile 97%

Dodavatel: Apollo Scientific

PC8433-1G PC8433-500MG PC8433-5G
APOSPC8433-1GEA 11540 CZK
APOSPC8433-1G APOSPC8433-500MG APOSPC8433-5G
3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}-5-methyl-4-isothiazolecarbonitrile 97%
3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}-5-methyl-4-isothiazolecarbonitrile
Vzorec: C₁₁H₅ClF₃N₃S₂
Bod tání: 122…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00244313
CAS číslo: 338775-63-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR