Tisk…

2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}acetohydrazide 97%

Dodavatel: Apollo Scientific

PC8430-1G PC8430-500MG
APOSPC8430-1GEA 22390 CZK
APOSPC8430-1G APOSPC8430-500MG
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}acetohydrazide 97%
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}acetohydrazide
Vzorec: C₈H₇ClF₃N₃OS
MW: 285,68 g/mol
Bod tání: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00664272
CAS číslo: 338422-76-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR