Tisk…

{4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl}acetonitrile 97%

Dodavatel: Apollo Scientific

PC8421-1G PC8421-500MG
APOSPC8421-1GEA 27840 CZK
APOSPC8421-1G APOSPC8421-500MG
{4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl}acetonitrile 97%
{4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl}acetonitrile
Vzorec: C₁₂H₁₂ClF₃N₄
Bod tání: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00141716
CAS číslo: 338979-38-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR