Tisk…

4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yloxy]phenyacetonitrile 97%

Dodavatel: Apollo Scientific

PC8418-1G PC8418-500MG
APOSPC8418-1GEA 27840 CZK
APOSPC8418-1G APOSPC8418-500MG
4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yloxy]phenyacetonitrile 97%
4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yloxy]phenyacetonitrile
MW: 312,68 g/mol
Bod tání: 86…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00172779
CAS číslo: 339029-95-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR