Tisk…

2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Fluoro-3-(trifluoromethyl)phenacyl bromide, 2-Bromo-1-(4-fluoro-3-(trifluoromethyl)phenyl)ethanone

PC8410-1G PC8410-25G PC8410-5G
APOSPC8410-1GEA 622 CZK
APOSPC8410-1G APOSPC8410-25G APOSPC8410-5G
2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone 95%
2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone
Vzorec: C₉H₅BrF₄O
MW: 285,0330 g/mol
Bod tání: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD03094509
CAS číslo: 537050-14-9
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR