Tisk…

4-Fluoro-3-iodo-1H-indazole 97%

Dodavatel: Apollo Scientific
Varování

PC8397-1G PC8397-250MG PC8397-5G
APOSPC8397-1GEA 1700 CZK
APOSPC8397-1G APOSPC8397-250MG APOSPC8397-5G
4-Fluoro-3-iodo-1H-indazole 97%
4-Fluoro-3-iodo-1H-indazole
Vzorec: C₇H₄FIN₂
MW: 262,0200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781590
CAS číslo: 518990-32-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR