Tisk…

2-Fluoro-4-(2-methylallyl)anisole

Dodavatel: Apollo Scientific

PC8394-1G PC8394-5G
APOSPC8394-1GEA 6820 CZK
APOSPC8394-1G APOSPC8394-5G
2-Fluoro-4-(2-methylallyl)anisole
2-Fluoro-4-(2-methylallyl)anisole
Vzorec: C₁₁H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD07775005
CAS číslo: 951888-11-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR