Tisk…

5-Chloro-2-fluoro-4-methylpyridine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-Chloro-2-fluoro-4-picoline

PC8385-5G PC8385-1G PC8385-25G
APOSPC8385-5GEA 3020 CZK
APOSPC8385-5G APOSPC8385-1G APOSPC8385-25G
5-Chloro-2-fluoro-4-methylpyridine
5-Chloro-2-fluoro-4-methylpyridine
Vzorec: C₆H₅ClFN
MW: 145,56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972410
CAS číslo: 884494-88-6
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR