6-fluoro-4-methylpyridin-3-amine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
6-Fluoro-4-methylpyridin-3-amine, 5-Amino-2-fluoro-4-methylpyridine
PC8384-1G
PC8384-25G
APOSPC8384-1GEA
992
CZK
APOSPC8384-1G
APOSPC8384-25G
6-fluoro-4-methylpyridin-3-amine 97%
6-fluoro-4-methylpyridin-3-amine
Vzorec:
C₆H₇FN₂ MW: 126.134 g/mol Bod tání: 57…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD09864696 CAS číslo: 954236-33-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...